missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chlorophenyl)piperazine monohydrochloride, 97%

$47.49 - $174.26

Chemical Identifiers

CAS 13078-12-1
Molecular Formula C10H14Cl2N2
Molecular Weight (g/mol) 233.136
MDL Number MFCD00060187
InChI Key ZHGRQBSZTVJDHU-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl piperazine hydrochloride, para-chlorophenylpiperazine hydrochloride, timtec-bb sbb003262, piperazine, 1-4-chlorophenyl-, monohydrochloride, 4-chlorophenyl piperazine, chloride, 1-4-chlorophenyl piperazine;hydrochloride, 1-4-chlorophenyl-piperazine hydrochloride, acmc-1c3h8, 1-4-chlorophenyl piperazine hcl, 1-4-chlorophenyl piperazinehydrochloride
PubChem CID 3084720
IUPAC Name 1-(4-chlorophenyl)piperazine;hydrochloride
SMILES C1CN(CCN1)C2=CC=C(C=C2)Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0171206 View Documents Alfa Aesar™
L0171206
5g Each for $47.49
 
AAL0171214 View Documents Alfa Aesar™
L0171214
25g Each for $174.26
 
Specifications

Chemical Identifiers

13078-12-1
233.136
ZHGRQBSZTVJDHU-UHFFFAOYSA-N
3084720
C1CN(CCN1)C2=CC=C(C=C2)Cl.Cl
C10H14Cl2N2
MFCD00060187
1-4-chlorophenyl piperazine hydrochloride, para-chlorophenylpiperazine hydrochloride, timtec-bb sbb003262, piperazine, 1-4-chlorophenyl-, monohydrochloride, 4-chlorophenyl piperazine, chloride, 1-4-chlorophenyl piperazine;hydrochloride, 1-4-chlorophenyl-piperazine hydrochloride, acmc-1c3h8, 1-4-chlorophenyl piperazine hcl, 1-4-chlorophenyl piperazinehydrochloride
1-(4-chlorophenyl)piperazine;hydrochloride

Specifications

~277°C
13078-12-1
MFCD00060187
1-4-chlorophenyl piperazine hydrochloride, para-chlorophenylpiperazine hydrochloride, timtec-bb sbb003262, piperazine, 1-4-chlorophenyl-, monohydrochloride, 4-chlorophenyl piperazine, chloride, 1-4-chlorophenyl piperazine;hydrochloride, 1-4-chlorophenyl-piperazine hydrochloride, acmc-1c3h8, 1-4-chlorophenyl piperazine hcl, 1-4-chlorophenyl piperazinehydrochloride
C1CN(CCN1)C2=CC=C(C=C2)Cl.Cl
233.136
233.14
1-(4-Chlorophenyl)piperazine monohydrochloride
5g
C10H14Cl2N2
5647914
ZHGRQBSZTVJDHU-UHFFFAOYSA-N
1-(4-chlorophenyl)piperazine;hydrochloride
3084720
97%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger