Please login to your online account to display your discounted pricing

1-(4-Chlorophenyl)piperazine, 97%, Maybridge

$44.04 - $44.04

Chemical Identifiers

Chemical Name or Material 1-(4-Chlorophenyl)piperazine dihydrochloride
CAS 38869-46-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 97%
Molecular Formula C10H13ClN2·2HCl
Formula Weight 269.59
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC109920050

  SDS
thermo fisher scientific
109920050
5g Each for $44.04
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 269.59
Melting Point 274° to 279°C (dec.)
Quantity 5g
Color Beige to slightly brown
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
CAS 38869-46-4
Assay Percent Range 97%
Chemical Name or Material 1-(4-Chlorophenyl)piperazine dihydrochloride
Molecular Formula C10H13ClN2·2HCl