Please login to your online account to display your discounted pricing

1-(4-Chlorophenyl)piperazine, 97%, Maybridge


Quantity 5g

Manufacturer: thermo fisher scientific  109920050

 View more versions of this product

Catalog No. AC109920050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥94%
Formula Weight 269.59
Melting Point 274° to 279°C (dec.)
Quantity 5g
Color Beige to slightly brown
CAS Min % 94.0
CAS Max % 100.0
CAS 38869-46-4
Infrared Spectrum Authentic
Chemical Name or Material 1-(4-Chlorophenyl)piperazine dihydrochloride
Assay Percent Range 97%
Molecular Formula C10H13ClN2·2HCl

Specifications

Chemical Name or Material 1-(4-Chlorophenyl)piperazine dihydrochloride
CAS 38869-46-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 97%
Molecular Formula C10H13ClN2·2HCl
Formula Weight 269.59