Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chlorophenyl)ethanol, 97%

$44.45 - $173.28

Chemical Identifiers

Chemical Name or Material 1-(4-Chlorophenyl)ethanol
CAS 3391-10-4
Molecular Formula C8H9ClO
Formula Weight 156.61
MDL Number MFCD00041038
Beilstein 2042027
Synonym 4-Chloro-alpha-methylbenzyl alcohol; 4-Chlorophenyl methyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0503814

  SDS
alfa aesar™
L0503814
25g Each for $173.28

AAL0503806

  SDS
alfa aesar™
L0503806
5g Each for $44.45
Description & Specifications

Specifications

Chemical Name or Material 1-(4-Chlorophenyl)ethanol
Flash Point >110°C (230°F)
Boiling Point 80° to 82°C (1mmHg)
Quantity 25g
CAS 3391-10-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 97%
Molecular Formula C8H9ClO
Formula Weight 156.61
MDL Number MFCD00041038
Beilstein 2042027
Synonym 4-Chloro-alpha-methylbenzyl alcohol; 4-Chlorophenyl methyl carbinol
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.