Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid, 95%

$82.97 - $347.88

Chemical Identifiers

Chemical Name or Material 1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid
CAS 58880-37-8
Molecular Formula C13H15ClO2
Formula Weight 238.71
MDL Number MFCD00019350
Beilstein 3297868
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0533606

  SDS
alfa aesar™
L0533606
5g Each for $82.97

AAL0533614

  SDS
alfa aesar™
L0533614
25g Each for $347.88
Description & Specifications

Specifications

Formula Weight 238.71
Chemical Name or Material 1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid
CAS 58880-37-8
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 95%
Molecular Formula C13H15ClO2
MDL Number MFCD00019350
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 151° to 153°C
Quantity 5g
Beilstein 3297868
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.