Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chlorophenyl)-2-(isopropylamino)ethanol, 98%

$77.59 - $283.27

Chemical Identifiers

Chemical Name or Material 1-(4-Chlorophenyl)-2-(isopropylamino)ethanol
CAS 23299-13-0
Molecular Formula C11H16ClNO
Formula Weight 213.71
MDL Number MFCD08060970
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5075603

  SDS
alfa aesar™
H5075603
1g Each for $283.27

AAH50756MD

  SDS
alfa aesar™
H50756MD
250mg Each for $77.59
Description & Specifications

Specifications

Formula Weight 213.71
Health Hazard 1 Warning
Assay 98%
Chemical Name or Material 1-(4-Chlorophenyl)-2-(isopropylamino)ethanol
Molecular Formula C11H16ClNO
CAS 23299-13-0
Quantity 1g
Melting Point 106° to 108°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD08060970
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.