Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol, 98%

Chemical Identifiers

Chemical Name or Material 1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol
CAS 133902-66-6
Formula Weight 209.63
MDL Number MFCD00140928
Beilstein 4250389
Synonym 1-(4-Chlorophenyl)-4-hydroxymethyl-1H-1,2,3-triazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6473703

  SDS
alfa aesar™
H6473703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6473706

  SDS
alfa aesar™
H6473706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64737MD

  SDS
alfa aesar™
H64737MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 141° to 145°C
Formula Weight 209.63
Quantity 1g
Synonym 1-(4-Chlorophenyl)-4-hydroxymethyl-1H-1,2,3-triazole
Beilstein 4250389
Assay 98%
Chemical Name or Material 1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol
MDL Number MFCD00140928
CAS 133902-66-6
Physical Form Solid