missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6473706
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol | |
141°C to 145°C | |
MFCD00140928 | |
4250389 | |
CAHIFLPAMJOAGI-UHFFFAOYSA-N | |
[1-(4-chlorophenyl)triazol-4-yl]methanol | |
2763878 | |
98% |
133902-66-6 | |
C9H8ClN3O | |
5 g | |
1-4-chlorophenyl-1h-1,2,3-triazol-4-yl methanol, 1-4-chlorophenyl triazol-4-yl methanol, 1-4-chlorophenyl-1,2,3-triazol-4-yl methanol, 1-4-chlorophenyl-1h-1,2,3-triazole-4-yl-methanol, 1-4-chlorophenyl-1,2,3-triazol-4-yl methan-1-ol, pubchem13003, chlorophenyltriazolylmethanol, 1-4-chlorophenyl-1h-1,2,3-triazol-4-ylmethanol | |
C1=CC(=CC=C1N2C=C(N=N2)CO)Cl | |
209.633 | |
209.63 | |
Solid |
Chemical Identifiers
133902-66-6 | |
209.633 | |
CAHIFLPAMJOAGI-UHFFFAOYSA-N | |
2763878 | |
C1=CC(=CC=C1N2C=C(N=N2)CO)Cl |
C9H8ClN3O | |
MFCD00140928 | |
1-4-chlorophenyl-1h-1,2,3-triazol-4-yl methanol, 1-4-chlorophenyl triazol-4-yl methanol, 1-4-chlorophenyl-1,2,3-triazol-4-yl methanol, 1-4-chlorophenyl-1h-1,2,3-triazole-4-yl-methanol, 1-4-chlorophenyl-1,2,3-triazol-4-yl methan-1-ol, pubchem13003, chlorophenyltriazolylmethanol, 1-4-chlorophenyl-1h-1,2,3-triazol-4-ylmethanol | |
[1-(4-chlorophenyl)triazol-4-yl]methanol |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures