Please login to your online account to display your discounted pricing

1-(4-Chlorophenyl)-1-cyclopropanecarbonitrile, 98%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  170600500

 View more versions of this product

Catalog No. AC170600500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 177.63
Physical Form Crystalline Powder, Crystals or Chunks
Quantity 50g
Color White
TSCA TSCA
EINECS Number 264-871-0
Melting Point 52° to 56°C
CAS Max % 100.0
Assay Percent Range 98%
MDL Number MFCD00019211
Name Note 98%
Linear Formula ClC6H4(C3H4)CN
CAS Min % 97.5
CAS 64399-27-5
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopropanecarbonitrile
Molecular Formula C10 H8 Cl N
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopropanecarbonitrile
Name Note 98%
CAS 64399-27-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10 H8 Cl N
Linear Formula ClC6H4(C3H4)CN
Formula Weight 177.63
MDL Number MFCD00019211
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents