Please login to your online account to display your discounted pricing

1-(4-Chlorophenyl)-1-cyclopropanecarbonitrile, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  170600100

 View more versions of this product

Catalog No. AC170600100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder, Crystals or Chunks
Formula Weight 177.63
Quantity 10g
Color White
TSCA TSCA
EINECS Number 264-871-0
Melting Point 52° to 56°C
CAS Max % 100.0
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopropanecarbonitrile
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C10 H8 Cl N
Linear Formula ClC6H4(C3H4)CN
MDL Number MFCD00019211
CAS 64399-27-5
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopropanecarbonitrile
Name Note 98%
CAS 64399-27-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10 H8 Cl N
Linear Formula ClC6H4(C3H4)CN
Formula Weight 177.63
MDL Number MFCD00019211
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents