Please login to your online account to display your discounted pricing

1-(4-Chlorophenyl)-1-cyclopropanecarbonitrile, 98%, ACROS Organics™

$214.62 - $214.62

Chemical Identifiers

Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopropanecarbonitrile
Name Note 98%
CAS 64399-27-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10 H8 Cl N
Linear Formula ClC6H4(C3H4)CN
Formula Weight 177.63
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC170600500

  SDS
thermo fisher scientific
170600500
Glass bottle 50g Each for $214.62

AC170600100

  SDS
fisher scientific acros
170600100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder, Crystals or Chunks
Formula Weight 177.63
Quantity 50g
Color White
EINECS Number 264-871-0
TSCA TSCA
Melting Point 52° to 56°C
CAS Max % 100.0
Assay Percent Range 98%
MDL Number MFCD00019211
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopropanecarbonitrile
Packaging Glass bottle
Name Note 98%
Molecular Formula C10 H8 Cl N
Linear Formula ClC6H4(C3H4)CN
CAS 64399-27-5
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.