missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 161570250
Specifications
1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid | |
97.5 | |
Authentic | |
Glass bottle | |
ClC6H4C5H8CO2H | |
25g | |
QJNFJEMGWIQMJT-UHFFFAOYSA-N | |
1-(4-chlorophenyl)cyclopentane-1-carboxylic acid | |
97447 | |
98% |
80789-69-1 | |
100.0 | |
98% | |
C12H13ClO2 | |
MFCD00001373 | |
1-4-chlorophenyl cyclopentanecarboxylic acid, 1-4-chlorophenyl-1-cyclopentanecarboxylic acid, 1-4-chlorophenyl cyclopentane-1-carboxylic acid, cyclopentanecarboxylic acid, 1-4-chlorophenyl, asischem d50961, timtec-bb sbb000649, acmc-209pk2, labotest-bb lt00453384, 1-4-chloro phenyl cyclopentanecarboxylic acid, 1-4-chloro-phenyl-cyclopentylcarboxylic acid | |
C1CCC(C1)(C2=CC=C(C=C2)Cl)C(=O)O | |
224.69 | |
224.69 |
Safety and Handling
EINECSNumber : 279-552-1
RUO â Research Use Only