missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Chlorophenyl)-1-cyclohexanecarboxylic acid, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 170760250

 View more versions of this product

Catalog No. AC170760250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94%
238.71
Light gray
100.0
58880-37-8
95%
MFCD00019350
95%
Glass bottle
Powder and Granules
25g
150° to 156°C
94.0
ClC6H4(C6H10)COOH
C13H15ClO2
1-(4-Chlorophenyl)-1-cyclohexanecarboxylic acid
Authentic

Chemical Identifiers

58880-37-8
100.0
C13H15ClO2
238.71
1-(4-Chlorophenyl)-1-cyclohexanecarboxylic acid, 95%
94.0
95%
ClC6H4(C6H10)COOH
MFCD00019350
≥94%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 261-481-2

TSCA : TSCA