missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Chlorophenyl)-1-cyclohexanecarboxylic acid, 95%, ACROS Organics™

Manufacturer:  Acros Organics 170760050

 View more versions of this product

Catalog No. AC170760050


 

Specifications

Specifications

1-(4-Chlorophenyl)-1-cyclohexanecarboxylic acid
150°C to 156°C
95%
94.0
Authentic
≥94%
C13H15ClO2
MFCD00019350
C1CCC(CC1)(C2=CC=C(C=C2)Cl)C(=O)O
238.711
238.71
Gray
5g
58880-37-8
100.0
Glass bottle
95%
ClC6H4(C6H10)COOH
UPNXUJXIIZGXLQ-UHFFFAOYSA-N
1-(4-chlorophenyl)cyclohexane-1-carboxylic acid
100873
Powder and Granules

Chemical Identifiers

58880-37-8
238.711
UPNXUJXIIZGXLQ-UHFFFAOYSA-N
C13H15ClO2
MFCD00019350
100873
Safety and Handling

Safety and Handling

EINECSNumber : 261-481-2

TSCA : TSCA

SDS Documents