missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Chlorophenyl)-1-cyclohexanecarboxylic acid, 95%, ACROS Organics™

$66.42 - $66.42

Chemical Identifiers

CAS 58880-37-8
Molecular Formula C13H15ClO2
Molecular Weight (g/mol) 238.711
MDL Number MFCD00019350
InChI Key UPNXUJXIIZGXLQ-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl cyclohexanecarboxylic acid, 1-4-chlorophenyl cyclohexane-1-carboxylic acid, 1-4-chlorophenyl-1-cyclohexanecarboxylic acid, 1-4-chlorophenyl-1-cyclohexane-carboxylic acid, cyclohexanecarboxylic acid, 1-4-chlorophenyl, acmc-1cuiq, 4-chlorophenylcyclohexanecarboxylic acid, 4-chlorophenyl cyclohexanecarboxylic acid, 1-4-chlorophenyl cyclohexanecarboxylicacid, 1-4-chlorophenyl cyclohexanecarboxylic acid
PubChem CID 100873
IUPAC Name 1-(4-chlorophenyl)cyclohexane-1-carboxylic acid
SMILES C1CCC(CC1)(C2=CC=C(C=C2)Cl)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC170760250 SDS View Documents Acros Organics
170760250
25g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC170760050 SDS View Documents Acros Organics
170760050
5g Glass bottle Each for $66.42
Specifications

Chemical Identifiers

58880-37-8
238.711
UPNXUJXIIZGXLQ-UHFFFAOYSA-N
100873
C1CCC(CC1)(C2=CC=C(C=C2)Cl)C(=O)O
C13H15ClO2
MFCD00019350
1-4-chlorophenyl cyclohexanecarboxylic acid, 1-4-chlorophenyl cyclohexane-1-carboxylic acid, 1-4-chlorophenyl-1-cyclohexanecarboxylic acid, 1-4-chlorophenyl-1-cyclohexane-carboxylic acid, cyclohexanecarboxylic acid, 1-4-chlorophenyl, acmc-1cuiq, 4-chlorophenylcyclohexanecarboxylic acid, 4-chlorophenyl cyclohexanecarboxylic acid, 1-4-chlorophenyl cyclohexanecarboxylicacid, 1-4-chlorophenyl cyclohexanecarboxylic acid
1-(4-chlorophenyl)cyclohexane-1-carboxylic acid

Specifications

Authentic
Gray
25g
94.0
95%
ClC6H4(C6H10)COOH
1-4-chlorophenyl cyclohexanecarboxylic acid, 1-4-chlorophenyl cyclohexane-1-carboxylic acid, 1-4-chlorophenyl-1-cyclohexanecarboxylic acid, 1-4-chlorophenyl-1-cyclohexane-carboxylic acid, cyclohexanecarboxylic acid, 1-4-chlorophenyl, acmc-1cuiq, 4-chlorophenylcyclohexanecarboxylic acid, 4-chlorophenyl cyclohexanecarboxylic acid, 1-4-chlorophenyl cyclohexanecarboxylicacid, 1-4-chlorophenyl cyclohexanecarboxylic acid
C1CCC(CC1)(C2=CC=C(C=C2)Cl)C(=O)O
238.711
238.71
≥94%
Glass bottle
150°C to 156°C
58880-37-8
100.0
C13H15ClO2
MFCD00019350
UPNXUJXIIZGXLQ-UHFFFAOYSA-N
1-(4-chlorophenyl)cyclohexane-1-carboxylic acid
100873
Powder and Granules
1-(4-Chlorophenyl)-1-cyclohexanecarboxylic acid, 95%
Safety and Handling

Safety and Handling

EINECSNumber : 261-481-2

TSCA : TSCA