Please login to your online account to display your discounted pricing

1-(4-Chlorophenyl)-1-cyclobutanecarboxylicacid 94%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC170721000

 View more versions of this product

Catalog No. AC170721000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 210.66
Physical Form Crystalline Powder
Assay ≥92% (HPLC)
Quantity 100g
Color Light yellow to purple
Melting Point 80° to 90°C
Molecular Formula C11H11ClO2
CAS 50921-39-6
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclobutanecarboxylic acid
Linear Formula ClC6H4C4H6CO2H

Specifications

Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclobutanecarboxylic acid
Assay ≥92% (HPLC)
Assay Percent Range 94%
Molecular Formula C11H11ClO2
Linear Formula ClC6H4C4H6CO2H
Formula Weight 210.66