Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chlorophenyl)-1-cyclobutanecarbonitrile, 97+%

Chemical Identifiers

Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclobutanecarbonitrile
CAS 28049-61-8
Molecular Formula C11H10ClN
Formula Weight 191.66
MDL Number MFCD00065239
UN Number UN3276
Synonym 1-(4-Chlorophenyl)-1-cyanocyclobutane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2778906

  SDS
alfa aesar™
H2778906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2778914

  SDS
alfa aesar™
H2778914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.66
Health Hazard 1 Danger
Assay ≥97%
Synonym 1-(4-Chlorophenyl)-1-cyanocyclobutane
Molecular Formula C11H10ClN
Boiling Point 295°C
MDL Number MFCD00065239
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
UN Number UN3276
CAS 28049-61-8
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclobutanecarbonitrile
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.