Please login to your online account to display your discounted pricing

1-(4-chlorophenethyl)-5-oxo-3-pyrrolidinecarboxylic acid, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO00129EA

 View more versions of this product

Catalog No. MO00129EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 267.71
Quantity 10g
Assay 97%
CAS Min % 97.0
CAS 368870-06-8
Molecular Formula C13H14ClNO3
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
MDL Number MFCD01330893
Packaging Amber Glass Bottle
Assay Percent Range 97%
Chemical Name or Material 1-(4-Chlorophenethyl)-5-oxo-3-pyrrolidinecarboxylic acid
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-(4-Chlorophenethyl)-5-oxo-3-pyrrolidinecarboxylic acid
CAS 368870-06-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H14ClNO3
Formula Weight 267.71
MDL Number MFCD01330893
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents