Please login to your online account to display your discounted pricing

1-(4-chlorophenethyl)-5-oxo-3-pyrrolidinecarboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-(4-Chlorophenethyl)-5-oxo-3-pyrrolidinecarboxylic acid
CAS 368870-06-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H14ClNO3
Formula Weight 267.71
MDL Number MFCD01330893
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00129DA

 
fisher scientific company
MO00129DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MO00129EA

 
fisher scientific company
MO00129EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 267.71
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 1-(4-Chlorophenethyl)-5-oxo-3-pyrrolidinecarboxylic acid
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C13H14ClNO3
MDL Number MFCD01330893
CAS 368870-06-8
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.