missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chlorobenzyl)piperazine, 98%

Manufacturer:  Alfa Aesar™ H5533714

 View more versions of this product

Catalog No. AAH5533714



Specifications

Specifications

1-(4-Chlorobenzyl)piperazine
>110°C (230°F)
104°C to 106°C (0.1mmHg)
25g
C11H15ClN2
UN3267
GSJXJZOWHSTWOX-UHFFFAOYSA-N
1-[(4-chlorophenyl)methyl]piperazine
134826
Liquid
1.140 g/mL
1.558
Colorless to Yellow
23145-88-2
MFCD00040791
1-4-chloro benzyl piperazine
C1CN(CCN1)CC2=CC=C(C=C2)Cl
210.705
210.71
98%

Chemical Identifiers

23145-88-2
210.705
GSJXJZOWHSTWOX-UHFFFAOYSA-N
134826
C1CN(CCN1)CC2=CC=C(C=C2)Cl
C11H15ClN2
MFCD00040791
1-4-chloro benzyl piperazine
1-[(4-chlorophenyl)methyl]piperazine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents