Learn More
1-(4-Chlorobenzyl)piperazine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5533703
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-(4-Chlorobenzyl)piperazine | |
Colorless to Yellow | |
104°C to 106°C (0.1 mmHg) | |
C11H15ClN2 | |
MFCD00040791 | |
UN3267 | |
GSJXJZOWHSTWOX-UHFFFAOYSA-N | |
1-[(4-chlorophenyl)methyl]piperazine | |
134826 | |
98% |
23145-88-2 | |
1.140 g/mL | |
>110°C (230°F) | |
1.558 | |
1 g | |
1-4-chlorobenzyl piperazine, 1-4-chlorophenyl methyl piperazine, 4-chlorobenzylpiperazine, 1-4-chlorobenzyl-piperazine, 1-4-chloro-benzyl-piperazine, 1-4-chloro benzyl piperazine, n-4-chlorobenzyl piperazine, 4-chlorophenyl methyl piperazine, norhomochlorcyclizine, chlorobenzylpiperazin | |
C1CN(CCN1)CC2=CC=C(C=C2)Cl | |
210.705 | |
210.71 | |
Liquid |
Chemical Identifiers
23145-88-2 | |
210.705 | |
GSJXJZOWHSTWOX-UHFFFAOYSA-N | |
134826 | |
C1CN(CCN1)CC2=CC=C(C=C2)Cl |
C11H15ClN2 | |
MFCD00040791 | |
1-4-chlorobenzyl piperazine, 1-4-chlorophenyl methyl piperazine, 4-chlorobenzylpiperazine, 1-4-chlorobenzyl-piperazine, 1-4-chloro-benzyl-piperazine, 1-4-chloro benzyl piperazine, n-4-chlorobenzyl piperazine, 4-chlorophenyl methyl piperazine, norhomochlorcyclizine, chlorobenzylpiperazin | |
1-[(4-chlorophenyl)methyl]piperazine |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only