missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chlorobenzhydryl)piperazine, tech. 90%

Manufacturer:  Alfa Aesar™ H55970MD

 View more versions of this product

Catalog No. AAH55970MD



Specifications

Specifications

1-(4-Chlorobenzhydryl)piperazine
178°C to 180°C (0.5mmHg)
250mg
C17H19ClN2
1-4-chlorobenzhydryl piperazine
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl
286.803
286.8
Technical
113°C (235°F)
65°C to 70°C
303-26-4
MFCD00005965
UZKBSZSTDQSMDR-UHFFFAOYSA-N
1-[(4-chlorophenyl)-phenylmethyl]piperazine
9340
90%

Chemical Identifiers

303-26-4
286.803
UZKBSZSTDQSMDR-UHFFFAOYSA-N
9340
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl
C17H19ClN2
MFCD00005965
1-4-chlorobenzhydryl piperazine
1-[(4-chlorophenyl)-phenylmethyl]piperazine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P260-P264b-P271-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c

H314-H317-H335-H361

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 206-137-4

RTECSNumber : TL1772000

TSCA : No

Recommended Storage : Ambient temperatures

Documents