Please login to your online account to display your discounted pricing

1-(4-Chlorobenzhydryl)piperazine, 90-95%, ACROS Organics™

$408.37 - $408.37

Chemical Identifiers

Chemical Name or Material 1-(4-Chlorobenzhydryl)piperazine
Assay ≥90% (on dry substance)
Assay Percent Range 90-95%
Molecular Formula C17H19ClN2
Formula Weight 286.79
Synonym 1-(4-Chlorophenylmethyl)piperazine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC108650050

  SDS
fisher scientific acros
108650050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC108650250

  SDS
thermo fisher scientific
108650250
Glass bottle 25g Each for $408.37

AC108651000

  SDS
thermo fisher scientific
AC108651000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 1-(4-Chlorobenzhydryl)piperazine
Color White to light beige
Melting Point 65° to 75°C
Quantity 5g
CAS 303-26-4
Infrared Spectrum Authentic
Packaging Glass bottle
Water 4% Max. (K.F.)
Physical Form Crystalline Powder
Assay ≥90% (on dry substance)
Assay Percent Range 90-95%
Molecular Formula C17H19ClN2
Formula Weight 286.79
Synonym 1-(4-Chlorophenylmethyl)piperazine