missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chloro-2-nitrophenyl)pyrrolidine, 97%

$35.93 - $422.54

Chemical Identifiers

CAS 41173-36-8
Molecular Formula C10H11ClN2O2
Molecular Weight (g/mol) 226.66
InChI Key QATZMIJXHKOELL-UHFFFAOYSA-N
Synonym 1-4-chloro-2-nitrophenyl pyrrolidine, 1-4-chloro-2-nitro-phenyl pyrrolidine, n-4-chloro-2-nitrophenyl pyrrolidine, 4-chloro-2-nitrophenyl pyrrolidine, qatzmijxhkoell-uhfffaoysa, 1-4-chloro-2-nitro-phenyl-pyrrolidine, 1-4-chloro-2-nitrophenyl pyrrolidine #, 1-4-chloranyl-2-nitro-phenyl pyrrolidine, 2-pyrrolidin-1-yl-5-chloro-1-nitrobenzene
PubChem CID 285321
IUPAC Name 1-(4-chloro-2-nitrophenyl)pyrrolidine
SMILES C1CCN(C1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63558MD View Documents Alfa Aesar™
H63558MD
250mg Each for $35.93
 
AAH6355803 View Documents Alfa Aesar™
H6355803
1g Each for $106.26
 
AAH6355806 View Documents Alfa Aesar™
H6355806
5g Each for $422.54
 
Specifications

Chemical Identifiers

41173-36-8
226.66
1-4-chloro-2-nitrophenyl pyrrolidine, 1-4-chloro-2-nitro-phenyl pyrrolidine, n-4-chloro-2-nitrophenyl pyrrolidine, 4-chloro-2-nitrophenyl pyrrolidine, qatzmijxhkoell-uhfffaoysa, 1-4-chloro-2-nitro-phenyl-pyrrolidine, 1-4-chloro-2-nitrophenyl pyrrolidine #, 1-4-chloranyl-2-nitro-phenyl pyrrolidine, 2-pyrrolidin-1-yl-5-chloro-1-nitrobenzene
1-(4-chloro-2-nitrophenyl)pyrrolidine
C10H11ClN2O2
QATZMIJXHKOELL-UHFFFAOYSA-N
285321
C1CCN(C1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]

Specifications

250mg
C10H11ClN2O2
1-4-chloro-2-nitrophenyl pyrrolidine, 1-4-chloro-2-nitro-phenyl pyrrolidine, n-4-chloro-2-nitrophenyl pyrrolidine, 4-chloro-2-nitrophenyl pyrrolidine, qatzmijxhkoell-uhfffaoysa, 1-4-chloro-2-nitro-phenyl-pyrrolidine, 1-4-chloro-2-nitrophenyl pyrrolidine #, 1-4-chloranyl-2-nitro-phenyl pyrrolidine, 2-pyrrolidin-1-yl-5-chloro-1-nitrobenzene
C1CCN(C1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
226.66
226.66
97%
41173-36-8
915044
QATZMIJXHKOELL-UHFFFAOYSA-N
1-(4-chloro-2-nitrophenyl)pyrrolidine
285321
Solid
1-(4-Chloro-2-nitrophenyl)pyrrolidine