missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chloro-2-nitrophenyl)piperidine, 97%

$35.93 - $422.54

Chemical Identifiers

CAS 33784-44-0
Molecular Formula C11H13ClN2O2
Molecular Weight (g/mol) 240.687
InChI Key YTFSYIPAIYQDPS-UHFFFAOYSA-N
Synonym 1-4-chloro-2-nitrophenyl piperidine, n-4-chloro-2-nitrophenyl piperidine, 4-chloro-2-nitrophenyl piperidine, 1-4-chloro-2-nitro-phenyl piperidine, 1-4-chloro-2-nitro-phenyl-piperidine, 1-4-chloro-2-nitrophenyl piperidine #, 1-4-chloranyl-2-nitro-phenyl piperidine
PubChem CID 285318
IUPAC Name 1-(4-chloro-2-nitrophenyl)piperidine
SMILES C1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63732MD View Documents Alfa Aesar™
H63732MD
250mg Each for $35.93
 
AAH6373203 View Documents Alfa Aesar™
H6373203
1g Each for $106.26
 
AAH6373206 View Documents Alfa Aesar™
H6373206
5g Each for $422.54
 
Specifications

Chemical Identifiers

33784-44-0
240.687
1-4-chloro-2-nitrophenyl piperidine, n-4-chloro-2-nitrophenyl piperidine, 4-chloro-2-nitrophenyl piperidine, 1-4-chloro-2-nitro-phenyl piperidine, 1-4-chloro-2-nitro-phenyl-piperidine, 1-4-chloro-2-nitrophenyl piperidine #, 1-4-chloranyl-2-nitro-phenyl piperidine
1-(4-chloro-2-nitrophenyl)piperidine
C11H13ClN2O2
YTFSYIPAIYQDPS-UHFFFAOYSA-N
285318
C1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]

Specifications

250mg
C11H13ClN2O2
1-4-chloro-2-nitrophenyl piperidine, n-4-chloro-2-nitrophenyl piperidine, 4-chloro-2-nitrophenyl piperidine, 1-4-chloro-2-nitro-phenyl piperidine, 1-4-chloro-2-nitro-phenyl-piperidine, 1-4-chloro-2-nitrophenyl piperidine #, 1-4-chloranyl-2-nitro-phenyl piperidine
C1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
240.687
240.69
97%
33784-44-0
203001
YTFSYIPAIYQDPS-UHFFFAOYSA-N
1-(4-chloro-2-nitrophenyl)piperidine
285318
Solid
1-(4-Chloro-2-nitrophenyl)piperidine