missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chloro-2-nitrophenyl)piperidine, 97%

Manufacturer:  Alfa Aesar™ H6373206

 View more versions of this product

Catalog No. AAH6373206



Specifications

Specifications

1-(4-Chloro-2-nitrophenyl)piperidine
33784-44-0
203001
YTFSYIPAIYQDPS-UHFFFAOYSA-N
1-(4-chloro-2-nitrophenyl)piperidine
285318
Solid
5g
C11H13ClN2O2
1-4-chloro-2-nitrophenyl piperidine, n-4-chloro-2-nitrophenyl piperidine, 4-chloro-2-nitrophenyl piperidine, 1-4-chloro-2-nitro-phenyl piperidine, 1-4-chloro-2-nitro-phenyl-piperidine, 1-4-chloro-2-nitrophenyl piperidine #, 1-4-chloranyl-2-nitro-phenyl piperidine
C1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
240.687
240.69
97%

Chemical Identifiers

33784-44-0
240.687
1-4-chloro-2-nitrophenyl piperidine, n-4-chloro-2-nitrophenyl piperidine, 4-chloro-2-nitrophenyl piperidine, 1-4-chloro-2-nitro-phenyl piperidine, 1-4-chloro-2-nitro-phenyl-piperidine, 1-4-chloro-2-nitrophenyl piperidine #, 1-4-chloranyl-2-nitro-phenyl piperidine
1-(4-chloro-2-nitrophenyl)piperidine
C11H13ClN2O2
YTFSYIPAIYQDPS-UHFFFAOYSA-N
285318
C1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
Documents