missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chloro-2-nitrophenyl)piperazine, 97%

$35.93 - $422.54

Chemical Identifiers

CAS 405910-34-1
Molecular Formula C10H12ClN3O2
Molecular Weight (g/mol) 241.675
MDL Number MFCD00640777
InChI Key NZFQOXSALNTRDH-UHFFFAOYSA-N
Synonym 1-4-chloro-2-nitrophenyl piperazine, 4-chloro-2-nitrophenyl piperazine, 1-4-chloro-2-nitro-phenyl-piperazine, piperazine, 1-4-chloro-2-nitrophenyl
PubChem CID 2735779
IUPAC Name 1-(4-chloro-2-nitrophenyl)piperazine
SMILES C1CN(CCN1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63851MD View Documents Alfa Aesar™
H63851MD
250mg Each for $35.93
 
AAH6385103 View Documents Alfa Aesar™
H6385103
1g Each for $106.26
 
AAH6385106 View Documents Alfa Aesar™
H6385106
5g Each for $422.54
 
Specifications

Chemical Identifiers

405910-34-1
241.675
NZFQOXSALNTRDH-UHFFFAOYSA-N
2735779
C1CN(CCN1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
C10H12ClN3O2
MFCD00640777
1-4-chloro-2-nitrophenyl piperazine, 4-chloro-2-nitrophenyl piperazine, 1-4-chloro-2-nitro-phenyl-piperazine, piperazine, 1-4-chloro-2-nitrophenyl
1-(4-chloro-2-nitrophenyl)piperazine

Specifications

250mg
C10H12ClN3O2
1-4-chloro-2-nitrophenyl piperazine, 4-chloro-2-nitrophenyl piperazine, 1-4-chloro-2-nitro-phenyl-piperazine, piperazine, 1-4-chloro-2-nitrophenyl
C1CN(CCN1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
241.675
241.67
97%
405910-34-1
MFCD00640777
NZFQOXSALNTRDH-UHFFFAOYSA-N
1-(4-chloro-2-nitrophenyl)piperazine
2735779
Solid
1-(4-Chloro-2-nitrophenyl)piperazine