missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chloro-2-nitrophenyl)homopiperazine, 96%

Manufacturer:  Alfa Aesar™ H6376603

 View more versions of this product

Catalog No. AAH6376603



Specifications

Specifications

1-(4-Chloro-2-nitrophenyl)homopiperazine
443923-21-5
1-(4-Chloro-2-nitrophenyl)-1,4-diazepane
Solid
1g
UN3259
255.7
96%

Chemical Identifiers

443923-21-5
1-(4-Chloro-2-nitrophenyl)-1,4-diazepane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

SDS Documents