Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chloro-2-nitrophenyl)-4-piperidone, 97%

Chemical Identifiers

Chemical Name or Material 1-(4-Chloro-2-nitrophenyl)-4-piperidone
Formula Weight 254.67
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6371703

 
alfa aesar™
H6371703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6371706

 
alfa aesar™
H6371706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63717MD

 
alfa aesar™
H63717MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 254.67
Quantity 1g
Chemical Name or Material 1-(4-Chloro-2-nitrophenyl)-4-piperidone
Assay 97%
Physical Form Solid