missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chloro-2-nitrophenyl)-4-methylpiperazine, 97%

$35.93 - $422.54

Chemical Identifiers

CAS 65639-68-1
Molecular Formula C11H14ClN3O2
Molecular Weight (g/mol) 255.702
InChI Key GYZYCENBDCHKGE-UHFFFAOYSA-N
Synonym 1-4-chloro-2-nitrophenyl-4-methylpiperazine, piperazine, 1-4-chloro-2-nitrophenyl-4-methyl, cambridge id 5670746, 1-4-chloro-2-nitro-phenyl-4-methyl-piperazine
PubChem CID 2860302
IUPAC Name 1-(4-chloro-2-nitrophenyl)-4-methylpiperazine
SMILES CN1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63734MD View Documents Alfa Aesar™
H63734MD
250mg Each for $35.93
 
AAH6373403 View Documents Alfa Aesar™
H6373403
1g Each for $106.26
 
AAH6373406 View Documents Alfa Aesar™
H6373406
5g Each for $422.54
 
Specifications

Chemical Identifiers

65639-68-1
255.702
1-4-chloro-2-nitrophenyl-4-methylpiperazine, piperazine, 1-4-chloro-2-nitrophenyl-4-methyl, cambridge id 5670746, 1-4-chloro-2-nitro-phenyl-4-methyl-piperazine
1-(4-chloro-2-nitrophenyl)-4-methylpiperazine
C11H14ClN3O2
GYZYCENBDCHKGE-UHFFFAOYSA-N
2860302
CN1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]

Specifications

250mg
C11H14ClN3O2
GYZYCENBDCHKGE-UHFFFAOYSA-N
1-(4-chloro-2-nitrophenyl)-4-methylpiperazine
2860302
Solid
1-(4-Chloro-2-nitrophenyl)-4-methylpiperazine
65639-68-1
1-4-chloro-2-nitrophenyl-4-methylpiperazine, piperazine, 1-4-chloro-2-nitrophenyl-4-methyl, cambridge id 5670746, 1-4-chloro-2-nitro-phenyl-4-methyl-piperazine
CN1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
255.702
255.7
97%