missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Butanediphosphonic acid, 98%, ACROS Organics™

$180.40 - $180.40

Chemical Identifiers

CAS 4671-77-6
Molecular Formula C4H12O6P2
Molecular Weight (g/mol) 218.082
MDL Number MFCD01631212
InChI Key JKTORXLUQLQJCM-UHFFFAOYSA-N
Synonym 1,4-butanediphosphonic acid, 1,4-butylenediphosphonic acid, 1,4-diphosphonobutane, butane-1,4-bisphosphonic acid, butane-1,4-diyldiphosphonic acid, tetramethylene phosphonic acid, tetramethylenebisphosphonic acid
PubChem CID 437082
IUPAC Name 4-phosphonobutylphosphonic acid
SMILES C(CCP(=O)(O)O)CP(=O)(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC332200010
SDS View Documents
Thermo Fisher Scientific
332200010
1g Glass Bottle
Each for $180.40
 
Description

Description

Specifications

Chemical Identifiers

4671-77-6
218.082
JKTORXLUQLQJCM-UHFFFAOYSA-N
437082
C(CCP(=O)(O)O)CP(=O)(O)O
C4H12O6P2
MFCD01631212
1,4-butanediphosphonic acid, 1,4-butylenediphosphonic acid, 1,4-diphosphonobutane, butane-1,4-bisphosphonic acid, butane-1,4-diyldiphosphonic acid, tetramethylene phosphonic acid, tetramethylenebisphosphonic acid
4-phosphonobutylphosphonic acid

Specifications

Authentic
White
1g
97.5
98%
(O)P(OH)2(CH2)4P(OH)2(O)
1,4-butanediphosphonic acid, 1,4-butylenediphosphonic acid, 1,4-diphosphonobutane, butane-1,4-bisphosphonic acid, butane-1,4-diyldiphosphonic acid, tetramethylene phosphonic acid, tetramethylenebisphosphonic acid
C(CCP(=O)(O)O)CP(=O)(O)O
218.082
218.08
98%
Glass Bottle
219.0°C to 221.0°C
4671-77-6
100.0
C4H12O6P2
MFCD01631212
JKTORXLUQLQJCM-UHFFFAOYSA-N
4-phosphonobutylphosphonic acid
437082
Fine Powder
1, 4-Butanediphosphonic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA

SDS