missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Butanediol diglycidyl ether, 96%

Manufacturer:  Alfa Aesar™ L1263209

 View more versions of this product

Catalog No. AAL1263209



Description

Description

Applications
1,4-Butanediol diglycidyl ether is used for preparing amylose, xylan and hydroxyethyl-cellulose, to cross link polyethylenimine which also serves as a hole-blocking layer in organic solar cells, to cross link hyaluronic acid into hydrogels and for the activation of soluble dextran polymers. It is the most commonly used homobifunctional epoxide compound

Solubility
Soluble in water (55,6 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store at room temperature.
Specifications

Specifications

1,4-Butanediol diglycidyl ether
157°C to 158°C (11mmHg)
2425-79-8
MFCD00005146
1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane
C1C(O1)COCCCCOCC2CO2
202.25
202.25
140°C (284°F)
10g
C10H18O4
115238
SHKUUQIDMUMQQK-UHFFFAOYSA-N
2-[4-(oxiran-2-ylmethoxy)butoxymethyl]oxirane
17046
96%

Chemical Identifiers

2425-79-8
202.25
SHKUUQIDMUMQQK-UHFFFAOYSA-N
17046
C1C(O1)COCCCCOCC2CO2
C10H18O4
MFCD00005146
1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane
2-[4-(oxiran-2-ylmethoxy)butoxymethyl]oxirane
Safety and Handling

Safety and Handling

GHS H Statement
H312-H332-H315-H319-H317
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : EJ5100000

TSCA : TSCA

Documents