missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Butanediol Bis(3-aminocrotonate), TCI America™

$30.56 - $184.86

Chemical Identifiers

CAS 14205-47-1
Molecular Formula C12H20N2O4
Molecular Weight (g/mol) 256.302
MDL Number MFCD00059862
InChI Key DFUXQARZEWMFLQ-FKJILZIQSA-N
Synonym 1,4-Butylene Bis(3-aminocrotonate)
PubChem CID 22186498
IUPAC Name 4-[(E)-3-aminobut-2-enoyl]oxybutyl (Z)-3-aminobut-2-enoate
SMILES CC(=CC(=O)OCCCCOC(=O)C=C(C)N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B106925G View Documents TCI America
B106925G
25g Each for $30.56
 
B1069500G View Documents TCI America
B1069500G
500g Each for $184.86
 
Specifications

Chemical Identifiers

14205-47-1
256.302
DFUXQARZEWMFLQ-FKJILZIQSA-N
22186498
CC(=CC(=O)OCCCCOC(=O)C=C(C)N)N
C12H20N2O4
MFCD00059862
1,4-Butylene Bis(3-aminocrotonate)
4-[(E)-3-aminobut-2-enoyl]oxybutyl (Z)-3-aminobut-2-enoate

Specifications

Yellow
25g
C12H20N2O4
1,4-Butylene Bis(3-aminocrotonate)
CC(=CC(=O)OCCCCOC(=O)C=C(C)N)N
256.302
256.30
≥96.0% (GC)
148°C
14205-47-1
MFCD00059862
DFUXQARZEWMFLQ-FKJILZIQSA-N
4-[(E)-3-aminobut-2-enoyl]oxybutyl (Z)-3-aminobut-2-enoate
22186498
Crystal-Powder at 20°C
1,4-Butanediol Bis(3-aminocrotonate)
Safety and Handling

Safety and Handling

TSCA : Yes