missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Butane sultone, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC107675000

 View more versions of this product

Catalog No. AC107675000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Liquid
136.17
Clear colorless to yellow
99.0
0.1% max. (K.F.)
4-Hydroxybutane-1-sulfonic acid δ-sultone
1, 4-Butane sultone
99+%
99+%
100ppm max.
1.4600 to 1.4640 (20°C, 589nm)
≥99.0%
500mL
100.0
19, V,1, 10
Glass bottle
Authentic
C4H8O3S
1633-83-6
MFCD00006584
4-hydoxybutanesulfonic acid: < 0.1%

Chemical Identifiers

1633-83-6
100.0
C4H8O3S
MFCD00006584
1, 4-Butane sultone, 99+%
99.0
99+%
136.17
4-Hydroxybutane-1-sulfonic acid δ-sultone
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Suspected of causing genetic defects if inhaled.

GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 216-647-9

RTECSNumber : RP4300000

TSCA : TSCA