missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Bromophenyl)pyrrolidine, 97%, Maybridge

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C10H12BrN
Formula Weight 226.12
Chemical Name or Material 1-(4-Bromophenyl)pyrrolidine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC30110EA SDS Fisher Scientific Company
CC30110EA
Amber Glass Bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC30110DA SDS Fisher Scientific Company
CC30110DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC30110CB SDS Fisher Scientific Company
CC30110CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
226.12
97%
C10H12BrN
1-(4-Bromophenyl)pyrrolidine

Specifications

226.12
22090-26-2
Amber Glass Bottle
1-(4-Bromophenyl)pyrrolidine
10g
C10H12BrN
97%
97%