Please login to your online account to display your discounted pricing

1-(4-Bromophenyl)pyrrolidine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC30110DA

 View more versions of this product

Catalog No. CC30110DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 226.12
Quantity 1g
Assay 97%
Molecular Formula C10H12BrN
CAS 22090-26-2
Packaging Amber Glass Bottle
Chemical Name or Material 1-(4-Bromophenyl)pyrrolidine
Assay Percent Range 97%

Specifications

Chemical Name or Material 1-(4-Bromophenyl)pyrrolidine
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H12BrN
Formula Weight 226.12