Please login to your online account to display your discounted pricing

1-(4-Bromophenyl)octane, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB09864EA

 View more versions of this product

Catalog No. BTB09864EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 269.22
Quantity 10g
Assay 97%
CAS Max % 100.0
Linear Formula BrC6H4(CH2)7CH3
Chemical Name or Material 1-(4-Bromophenyl)octane
Assay Percent Range 97%
Molecular Formula C14H21Br
MDL Number MFCD00038348
CAS Min % 96.5
Packaging Amber Glass Bottle
CAS 51554-93-9

Specifications

Chemical Name or Material 1-(4-Bromophenyl)octane
CAS 51554-93-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H21Br
Linear Formula BrC6H4(CH2)7CH3
Formula Weight 269.22
MDL Number MFCD00038348