missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Bromophenyl)hexane, 97%, Thermo Scientific™

$16.45 - $107.58

Chemical Identifiers

CAS 23703-22-2
Molecular Formula C12H17Br
Molecular Weight (g/mol) 241.17
MDL Number MFCD00061114
InChI Key MXHOLIARBWJKCR-UHFFFAOYSA-N
Synonym 1-4-bromophenyl hexane, 4-hexylbromobenzene, 1-bromo-4-n-hexylbenzene, 4-bromo-1-hexylbenzene, benzene, 1-bromo-4-hexyl, 4-bromo-n-hexylbenzene, 4-hexylphenyl bromide, 1-bromo-4-hexyl-benzene, acmc-1cb1g, #
PubChem CID 598186
IUPAC Name 1-bromo-4-hexylbenzene
SMILES CCCCCCC1=CC=C(Br)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BTB09866DA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
BTB09866DA
1 g
Each for $16.45
 
BTB09866EA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
BTB09866EA
10 g
Each for $107.58
 
Description

Description

Specifications

Chemical Identifiers

23703-22-2
241.17
MXHOLIARBWJKCR-UHFFFAOYSA-N
598186
CCCCCCC1=CC=C(Br)C=C1
C12H17Br
MFCD00061114
1-4-bromophenyl hexane, 4-hexylbromobenzene, 1-bromo-4-n-hexylbenzene, 4-bromo-1-hexylbenzene, benzene, 1-bromo-4-hexyl, 4-bromo-n-hexylbenzene, 4-hexylphenyl bromide, 1-bromo-4-hexyl-benzene, acmc-1cb1g, #
1-bromo-4-hexylbenzene

Specifications

23703-22-2
100.0
Amber Glass Bottle
BrC6H4(CH2)5CH3
1 g
MXHOLIARBWJKCR-UHFFFAOYSA-N
1-bromo-4-hexylbenzene
598186
97%
96.5
97%
C12H17Br
MFCD00061114
1-4-bromophenyl hexane, 4-hexylbromobenzene, 1-bromo-4-n-hexylbenzene, 4-bromo-1-hexylbenzene, benzene, 1-bromo-4-hexyl, 4-bromo-n-hexylbenzene, 4-hexylphenyl bromide, 1-bromo-4-hexyl-benzene, acmc-1cb1g, #
CCCCCCC1=CC=C(Br)C=C1
241.17
241.17
1-(4-Bromophenyl)hexane
SDS
missing translation for 'documents'

missing translation for 'documents'