Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-1-(4-Bromophenyl)ethyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material (+/-)-1-(4-Bromophenyl)ethyl isocyanate
CAS 207974-15-0
Molecular Formula C9H8BrNO
Formula Weight 226.08
MDL Number MFCD00041411
UN Number UN2206
Synonym 4-Bromo-alpha-methylbenzyl isocyanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1183603

 
alfa aesar™
L1183603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1183606

 
alfa aesar™
L1183606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.08
Chemical Name or Material (+/-)-1-(4-Bromophenyl)ethyl isocyanate
Assay 97%
Molecular Formula C9H8BrNO
MDL Number MFCD00041411
CAS 207974-15-0
UN Number UN2206
Synonym 4-Bromo-alpha-methylbenzyl isocyanate
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage 2° to 8°C
Boiling Point 145° to 147°C (7mmHg)
Health Hazard 3 GHS P Statement
P284a-P261-P280h-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280h-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C