missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-1-(4-Bromophenyl)ethyl isocyanate, 97%

Manufacturer:  Alfa Aesar™ L1183606

 View more versions of this product

Catalog No. AAL1183606



Specifications

Specifications

(+/-)-1-(4-Bromophenyl)ethyl isocyanate
5g
C9H8BrNO
UN2206
AKVAGWOVWQDTAF-UHFFFAOYSA-N
1-bromo-4-(1-isocyanatoethyl)benzene
610983
97%
145°C to 147°C (7mmHg)
207974-15-0
MFCD00041411
1-bromo-4-1-isocyanatoethyl benzene, +/--1-4-bromophenyl ethyl isocyanate, acmc-20apkr, s-1-4-bromophenyl ethyl isocyanate, acmc-20api1, acmc-1c9h0, 1-4-bromophenyl ethanisocyanate, 1-4-bromophenyl ethyl isocyanate, 1-bromanyl-4-1-isocyanatoethyl benzene, 1-bromo-4-1-isocyanato-ethyl-benzene
CC(C1=CC=C(C=C1)Br)N=C=O
226.073
226.08

Chemical Identifiers

207974-15-0
226.073
AKVAGWOVWQDTAF-UHFFFAOYSA-N
610983
CC(C1=CC=C(C=C1)Br)N=C=O
C9H8BrNO
MFCD00041411
1-bromo-4-1-isocyanatoethyl benzene, +/--1-4-bromophenyl ethyl isocyanate, acmc-20apkr, s-1-4-bromophenyl ethyl isocyanate, acmc-20api1, acmc-1c9h0, 1-4-bromophenyl ethanisocyanate, 1-4-bromophenyl ethyl isocyanate, 1-bromanyl-4-1-isocyanatoethyl benzene, 1-bromo-4-1-isocyanato-ethyl-benzene
1-bromo-4-(1-isocyanatoethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P284a-P261-P280h-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents