Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Bromophenyl)ethanol, 95%

$28.01 - $218.58

Chemical Identifiers

Chemical Name or Material 1-(4-Bromophenyl)ethanol
CAS 5391-88-8
Molecular Formula C8H9BrO
Formula Weight 201.06
MDL Number MFCD00004514
Beilstein 2042029
Synonym 4-Bromo-alpha-methylbenzyl alcohol; 4-Bromophenyl methyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2356106

 
alfa aesar™
B2356106
5g Each for $28.01

AAB2356114

 
alfa aesar™
B2356114
25g Each for $68.35

AAB2356122

 
alfa aesar™
B2356122
100g Each for $218.58
Description & Specifications

Specifications

Formula Weight 201.06
Chemical Name or Material 1-(4-Bromophenyl)ethanol
Assay 95%
Molecular Formula C8H9BrO
MDL Number MFCD00004514
CAS 5391-88-8
Melting Point 36° to 38°C
Beilstein 2042029
Synonym 4-Bromo-alpha-methylbenzyl alcohol; 4-Bromophenyl methyl carbinol
Boiling Point 119° to 121°C (7mmHg)
Quantity 5g
Flash Point 63°C (145°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.