missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Bromophenyl)-2-phenylbenzimidazole, TCI America™

$168.06 - $574.45

Chemical Identifiers

CAS 760212-58-6
Molecular Formula C19H13BrN2
Molecular Weight (g/mol) 349.231
MDL Number MFCD09746342
InChI Key PPYIZNYOMNYZCG-UHFFFAOYSA-N
PubChem CID 23094071
IUPAC Name 1-(4-bromophenyl)-2-phenylbenzimidazole
SMILES C1=CC=C(C=C1)C2=NC3=CC=CC=C3N2C4=CC=C(C=C4)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B49291G View Documents TCI America
B49291G
1g Each for $168.06
 
B49295G View Documents TCI America
B49295G
5g Each for $574.45
 
Specifications

Chemical Identifiers

760212-58-6
349.231
PPYIZNYOMNYZCG-UHFFFAOYSA-N
1-(4-bromophenyl)-2-phenylbenzimidazole
C19H13BrN2
MFCD09746342
23094071
C1=CC=C(C=C1)C2=NC3=CC=CC=C3N2C4=CC=C(C=C4)Br

Specifications

Undesignated
1g
C19H13BrN2
PPYIZNYOMNYZCG-UHFFFAOYSA-N
1-(4-bromophenyl)-2-phenylbenzimidazole
23094071
Crystal-Powder at 20°C
1-(4-Bromophenyl)-2-phenylbenzimidazole
137°C
760212-58-6
MFCD09746342
C1=CC=C(C=C1)C2=NC3=CC=CC=C3N2C4=CC=C(C=C4)Br
349.231
349.23
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No