missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Bromophenyl)-2,5-dimethylpyrrole

$275.53 - $1,237.58

Chemical Identifiers

CAS 5044-24-6
Molecular Formula C12H12BrN
Formula Weight 250.14
MDL Number MFCD00277042
Chemical Name or Material 1-(4-Bromophenyl)-2,5-dimethylpyrrole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5059403 SDS Alfa Aesar™
H5059403
1g Each for $275.53
AAH5059406 SDS Alfa Aesar™
H5059406
5g Each for $1,237.58
Specifications Description & Specifications

Chemical Identifiers

5044-24-6
250.14
1-(4-Bromophenyl)-2,5-dimethylpyrrole
C12H12BrN
MFCD00277042

Specifications

5044-24-6
250.14
1g
C12H12BrN
MFCD00277042
1-(4-Bromophenyl)-2,5-dimethylpyrrole