Please login to your online account to display your discounted pricing

1-(4-Bromophenoxy)hexane, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  BTB09795DA

 View more versions of this product

Catalog No. BTB09795DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 257.17
Quantity 1g
Assay Percent Range 97%
Molecular Formula C12H17BrO
Chemical Name or Material 1-(4-Bromophenoxy)hexane
CAS 30752-19-3

Specifications

Chemical Name or Material 1-(4-Bromophenoxy)hexane
Assay Percent Range 97%
Molecular Formula C12H17BrO
Formula Weight 257.17