Please login to your online account to display your discounted pricing

1-(4-Bromophenoxy)hexane, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-(4-Bromophenoxy)hexane
Assay Percent Range 97%
Molecular Formula C12H17BrO
Formula Weight 257.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB09795DA

  SDS
fisher scientific company
BTB09795DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB09795EE

  SDS
fisher scientific company
BTB09795EE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB09795EA

  SDS
fisher scientific company
BTB09795EA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 257.17
Quantity 1g
Assay Percent Range 97%
Chemical Name or Material 1-(4-Bromophenoxy)hexane
Molecular Formula C12H17BrO
CAS 30752-19-3