Please login to your online account to display your discounted pricing

1-[4-(Bromomethyl)phenyl]-1H-pyrazole, 95%, Maybridge

$128.44 - $201.23

Chemical Identifiers

Chemical Name or Material 1-[4-(Bromomethyl)phenyl]-1H-pyrazole
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H9BrN2
Formula Weight 237.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC18408DE

 
fisher scientific company
CC18408DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC18408DA

  SDS
fisher scientific company
CC18408DA
Amber Glass Bottle 1g Each for $201.23

CC18408CB

  SDS
fisher scientific company
CC18408CB
Amber Glass Bottle 250mg Each for $128.44
Description & Specifications

Specifications

Formula Weight 237.1
Quantity 5g
Assay 95%
Assay Percent Range 95%
CAS 368869-85-6
Packaging Amber Glass Bottle
Chemical Name or Material 1-[4-(Bromomethyl)phenyl]-1H-pyrazole
Molecular Formula C10H9BrN2