Please login to your online account to display your discounted pricing

1-[4-(Bromomethyl)phenyl]-1H-pyrazole, 95%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC18408DA

 View more versions of this product

Catalog No. CC18408DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 237.1
Quantity 1g
Assay 95%
Chemical Name or Material 1-[4-(Bromomethyl)phenyl]-1H-pyrazole
Assay Percent Range 95%
Molecular Formula C10H9BrN2
CAS 368869-85-6
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 1-[4-(Bromomethyl)phenyl]-1H-pyrazole
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H9BrN2
Formula Weight 237.1