missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[4-(Bromomethyl)phenyl]-1H-1,2,4-triazole, 97%, Maybridge

Manufacturer:  Fisher Scientific CC18608DA

 View more versions of this product

Catalog No. CC18608DA



Specifications

Specifications

1-[4-(Bromomethyl)phenyl]-1H-1, 2, 4-triazole hydrobromide hydrate
1g
96.5
97%
MFCD04113600
NOUAEJFAVOHNSY-UHFFFAOYSA-N
1-[4-(bromomethyl)phenyl]-1,2,4-triazole
2776489
97%
Amber Glass Bottle
58419-69-5
100.0
C9H8BrN3
1-4-bromomethyl phenyl-1h-1,2,4-triazole, 1-4-bromomethyl phenyl-1h-1,2,4-triazole, 1-4-bromomethyl phenyl-1,2,4-triazole, 1h-1,2,4-triazole, 1-4-bromomethyl phenyl, 1-4-bromomethylphenyl-1h-1,2,4-triazole, zlchem 1040, 1-4-bromomethyl phenyl-1h-1,2,4-triazol, 1-4-bromomethyl-phenyl-1h-1,2,4-triazole, 1-4-bromo-methyl phenyl-1h-1,2,4-triazole, 1-4-bromomethyl-phenyl-1h-1,2,4-triazole
C1=CC(=CC=C1CBr)N2C=NC=N2
238.088
319

Chemical Identifiers

58419-69-5
238.088
NOUAEJFAVOHNSY-UHFFFAOYSA-N
2776489
C1=CC(=CC=C1CBr)N2C=NC=N2
C9H8BrN3
MFCD04113600
1-4-bromomethyl phenyl-1h-1,2,4-triazole, 1-4-bromomethyl phenyl-1h-1,2,4-triazole, 1-4-bromomethyl phenyl-1,2,4-triazole, 1h-1,2,4-triazole, 1-4-bromomethyl phenyl, 1-4-bromomethylphenyl-1h-1,2,4-triazole, zlchem 1040, 1-4-bromomethyl phenyl-1h-1,2,4-triazol, 1-4-bromomethyl-phenyl-1h-1,2,4-triazole, 1-4-bromo-methyl phenyl-1h-1,2,4-triazole, 1-4-bromomethyl-phenyl-1h-1,2,4-triazole
1-[4-(bromomethyl)phenyl]-1,2,4-triazole
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents