Please login to your online account to display your discounted pricing

1-(4-bromobutoxy)-3-nitrobenzene, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-(4-Bromobutoxy)-3-nitrobenzene
CAS 31191-44-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10 H12 Br N O3
Linear Formula NO2C6H4O(CH2)4Br
Formula Weight 274.11
MDL Number MFCD07368655
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC385280250

  SDS
thermo fisher scientific
AC385280250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Low Melting Solid
Formula Weight 274.11
Quantity 25g
Color Slightly yellow
Melting Point 32° to 35°C
CAS Min % 96.0
CAS Max % 100.0
CAS 31191-44-3
Linear Formula NO2C6H4O(CH2)4Br
Chemical Name or Material 1-(4-Bromobutoxy)-3-nitrobenzene
MDL Number MFCD07368655
Assay Percent Range 97%
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C10 H12 Br N O3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.