Learn More
1-(4-Bromobenzyl)piperazine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H55145MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-(4-Bromobenzyl)piperazine | |
56°C to 62°C | |
112°C to 116°C (0.3 mmHg) | |
MFCD02177416 | |
UN3259 | |
MAHWBNAOEVAPJF-UHFFFAOYSA-N | |
1-[(4-bromophenyl)methyl]piperazine | |
876494 | |
97% |
91345-62-9 | |
Brown to White | |
C11H15BrN2 | |
250 mg | |
1-4-bromobenzyl piperazine, 1-4-bromophenyl methyl piperazine, 1-4-bromo-benzyl-piperazine, piperazine, 1-4-bromophenyl methyl, cbmicro_035222, acmc-20apb8, cambridge id 5862395, 1-4-bromobenzyl-piperazine, 4-bromophenyl methyl piperazine | |
C1CN(CCN1)CC2=CC=C(C=C2)Br | |
255.159 | |
255.16 | |
Solid |
Chemical Identifiers
91345-62-9 | |
255.159 | |
MAHWBNAOEVAPJF-UHFFFAOYSA-N | |
876494 | |
C1CN(CCN1)CC2=CC=C(C=C2)Br |
C11H15BrN2 | |
MFCD02177416 | |
1-4-bromobenzyl piperazine, 1-4-bromophenyl methyl piperazine, 1-4-bromo-benzyl-piperazine, piperazine, 1-4-bromophenyl methyl, cbmicro_035222, acmc-20apb8, cambridge id 5862395, 1-4-bromobenzyl-piperazine, 4-bromophenyl methyl piperazine | |
1-[(4-bromophenyl)methyl]piperazine |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures